Learn More
bcl-x Antibody (5E9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26480-100ul
 View more versions of this product
Product Code. 30025849
  • 533.00 € / 100µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
bcl-x Monoclonal antibody specifically detects bcl-x in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
bcl-x
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, ELISA, Immunohistochemistry
5E9
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Apoptosis regulator Bcl-X,BCL2L,BCL2L1,Bcl-2-like protein 1,BCLX,Bcl-X,BCL-XL/S,PPP1R52
A synthesized peptide derived from Human Bcl-x [UniProt Q07817]
100 μL
Apoptosis, Biologically Active Proteins, Cancer, Phospho Specific, Tumor Suppressors
598
Store at -20 to -70C. Avoid freeze-thaw cycles.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications