missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bcl-rambo Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583173
This item is not returnable.
View return policy
Description
Immunogen sequence: QFGVTYLEDY SAEYIIQQGG WGTVFSLESE EEEYPGITAE DSNDIYILPS DNSGQVSPPE SPTVTTSWQS E Highest antigen sequence indentity to the following orthologs: Mouse - 89%, Rat - 90%.
Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer. Members of the Bcl-2 family are known to be critical regulators of this process. These proteins are characterized by the presence of several conserved motifs termed Bcl-2 homology (BH) domains. A novel, widely expressed member termed Bcl-rambo was recently identified. This protein is localized to mitochondria in mammalian cells and its overexpression induces apoptosis which could be blocked by co-expression of inhibitor of apoptosis proteins (IAPs) such as XIAP, cIAP1, and cIAP2. Bcl-rambo shows overall homology to the anti-apoptotic members containing BH motifs, but unlike Bcl-2, the C-terminal membrane anchor of Bcl-rambo is preceded by a unique 250 amino acid insertion. This region by itself can induce apoptosis more efficiently than the Bcl-2 homology regions, suggesting that Bcl-rambo may be important other pro-apoptotic pathways.
Specifications
| Bcl-rambo | |
| Polyclonal | |
| Unconjugated | |
| BCL2L13 | |
| BCL2 like 13; BCL2L13; Bcl2-L-13; BCL2-like 13 (apoptosis facilitator); bcl-2-like protein 13; Bcl-rambo; CD003; E430016C20Rik; MIL1; Mil-1; mitochondria located 1 homolog; protein Mil1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23786 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BXK5 | |
| BCL2L13 | |
| Recombinant protein corresponding to Human BCL2L13. Recombinant protein control fragment (Product #RP-95343). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction