missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-2 (Apoptosis & Follicular Lymphoma Marker) Mouse, Clone: rBCL2/782, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 596-MSM11-P1
This item is not returnable.
View return policy
Specifications
| Bcl-2 (Apoptosis & Follicular Lymphoma Marker) | |
| Outer mitochondrial membranes and endoplasmic reticulum as well as nuclear membranes. | |
| rBCL2/782 | |
| Unconjugated | |
| Mouse | |
| 25-26kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 596 | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Western Blot, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant full-length human bcl-2 protein | |
| Jurkat, K562, HL-60, or HeLa Cells. Tonsil or follicular lymphomas. | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction