missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAT1/ECA39 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13640-1-AP-150UL
This item is not returnable.
View return policy
Description
13640-1-AP antibody detects the bands around 43-45 kDa for different isoforms and higher molecular weight for phosphorylation modification of this protein.
This gene encodes the cytosolic form of the enzyme branched-chain amino acid transaminase. This enzyme catalyzes the reversible transamination of branched-chain alpha-keto acids to branched-chain L-amino acids essential for cell growth. Two different clinical disorders have been attributed to a defect of branched-chain amino acid transamination: hypervalinemia and hyperleucine-isoleucinemia. As there is also a gene encoding a mitochondrial form of this enzyme, mutations in either gene may contribute to these disorders. Alternatively spliced transcript variants have been described.Specifications
| BCAT1/ECA39 | |
| Polyclonal | |
| Unconjugated | |
| BCAT1 | |
| BCAT1, ECA39, MECA39, PNAS 121, PP18, Protein ECA39 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12035, 29592, 586 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P24288, P54687, P54690 | |
| Bcat1 | |
| BCAT1/ECA39 Fusion Protein Ag4574 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction