missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ IC-XT Pack CDC Anaerobe Agar Base with Penase

Product Code. 15565515
Change view
Click to view available options
:
CDC Anaerobe Agar; Quantity: 100; Diameter: 90 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15565515 CDC Anaerobe Agar; Quantity: 100; Diameter: 90 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15565515 Supplier BD Supplier No. 257614

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

BD Difco™ X100 CDC irradiated agar for environmental monitoring and detection of anaerobes. 90 mm diameter.

  • BD BBL™ IC-XT Pack CDC Anaerobe Agar Base with Penase
  • Used for the detection and enumeration of microorganism particularly obligate anaerobes in pharmaceutical clean rooms and production environments where penicillins are filled or processed.
  • All IC-XT products are packaged in impermeable plastic films to allow an extended stability and storage at 2°C to 25°C throughout the shelf life.
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Media Type CDC Anaerobe Agar
No. per Pack 100
Packaging Type Plates
Diameter (Metric) 90 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.