missing translation for 'onlineSavingsMsg'
Learn More

BAY 61-3606 dihydrochloride, MedChemExpress

Product Code. 16476278
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16476278

Brand: Medchem Express HY14985

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BAY 61-3606 dihydrochloride is an orally available, ATP-competitive, reversible and highly selective Syk inhibitor with a Ki of 7.5 nM an IC50 of 10 nM.

BAY 61-3606 dihydrochloride reduces ERK1/2 and Akt phosphorylation in neuroblastoma cell. BAY 61-3606 dihydrochloride induces a large decrease of Syk phosphorylation in K-rn cell lysates. Bay 61-3606 dihydrochloride sensitizes TRAIL-induced apoptosis by downregulating Mcl-1 in breast cancer cells.

Specifications

Chemical Name or Material BAY 61-3606 dihydrochloride
CAS 648903-57-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H20Cl2N6O3
Quantity 10 mg
Solubility Information H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(N)C1=CC=CN=C1NC2=NC(C3=CC=C(C(OC)=C3)OC)=CC4=NC=CN24.[H]Cl.[H]Cl
Molecular Weight (g/mol) 463.32
Formula Weight 463.32
Percent Purity 98.21%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.