missing translation for 'onlineSavingsMsg'
Learn More

Bay 60-7550, MedChemExpress

Product Code. 16468867
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468867 5 mg 5mg
16476616 100 mg 100mg
16406288 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16468867 Supplier Medchem Express Supplier No. HY14992

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bay 60-7550 is a potent and selective PDE2 inhibitor with a Ki of 3.8 nM.

Bay 60-7550 is a potent and selective PDE2 inhibitor with a Ki of 3.8 nM.

Specifications

Chemical Name or Material Bay 60-7550
CAS 439083-90-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H32N4O4
Quantity 5 mg
Synonym BAY 607550
Solubility Information DMSO : ≥ 33.3 mg/mL (69.87 mM)
SMILES O=C1N=C(NN2C1=C(N=C2[C@@H](CCCC3=CC=CC=C3)[C@@H](C)O)C)CC4=CC=C(C(OC)=C4)OC
Molecular Weight (g/mol) 476.57
Formula Weight 476.57
Percent Purity 98.12%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.