Learn More
Bay 60-7550, MedChemExpress
Product Code. 16476616
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476616 100 mg 100mg
16468867 5 mg 5mg
16406288 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16476616 Supplier Medchem Express Supplier No. HY14992
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Bay 60-7550 is a potent and selective PDE2 inhibitor with a Ki of 3.8 nM.
Bay 60-7550 is a potent and selective PDE2 inhibitor with a Ki of 3.8 nM.
Specifications
Chemical Name or Material Bay 60-7550
CAS 439083-90-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H32N4O4
Quantity 100 mg
Synonym BAY 607550
Solubility Information DMSO : ≥ 33.3 mg/mL (69.87 mM)
SMILES O=C1N=C(NN2C1=C(N=C2[C@@H](CCCC3=CC=CC=C3)[C@@H](C)O)C)CC4=CC=C(C(OC)=C4)OC
Molecular Weight (g/mol) 476.57
Formula Weight 476.57
Percent Purity 98.12%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration