missing translation for 'onlineSavingsMsg'
Learn More

BAY 11-7082, MedChemExpress

Product Code. 16425698
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16425698

Brand: Medchem Express HY13453

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BAY 11-7082 is an IκBα phosphorylation and NF-κB inhibitor.

BAY 11-7082 selectively and irreversibly inhibits the TNF-α-induced phosphorylation of IκB-α, and decreases NF-κB and expression of adhesion molecules. BAY 11-7082 inhibits ubiquitin-specific protease USP7 and USP21 (IC50=0.19, 0.96 μM, respectively). BAY 11-7082 inhibits gasdermin D (GSDMD) pore formation in liposomes and inflammasome-mediated pyroptosis and IL-1β secretion in human and mouse cells.

Specifications

Chemical Name or Material BAY 11-7082
CAS 19542-67-7
Color White
Purity Grade Notes Research
Molecular Formula C10H9NO2S
Quantity 10 mg
Synonym BAY 11-7821
Solubility Information DMSO : ≥ 100 mg/mL (482.51 mM)
SMILES N#C/C=C/S(=O)(C1=CC=C(C)C=C1)=O
Molecular Weight (g/mol) 207.25
Formula Weight 207.25
Percent Purity 99.28%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.