Learn More
Invitrogen™ Bax Chimeric Recombinant Rabbit Monoclonal Antibody (6A7)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547766
Description
Specificity: In the presence of nonionic detergent, the antibody avidly binds the monomeric form of Bax, but not Bax complexed with either Bcl-XL or Bcl-2. Bax antibody only recognizes conformationally changed active Bax. Preparation: KLH conjugated to peptides corresponding to amino acids 3-16 of murine Bax (CGSGEQLGSGGPTSS), amino acids 3-16 of human Bax (CGSGEQPRGGGPTSS), and amino acids 12-24 shared by Bax from both species (CGPTSSEQIMKTGA).
BAX is a members of the Bcl-2 Family and plays an important role in regulation of apoptosis. Whereas Bcl-2 is commonly regarded as an anti-apoptotic protein, BAX is considered to have a pro-apoptotic function. Regulation of apoptosis is supposed to involve both homo- and heterodimerization of different isoforms of BAX and Bcl-2. The Bax gene encodes different isoforms including Bax alpha (21 kDa) and Bax beta (24 kDa), whereas both isoforms contain the BH1, BH2 and BH3 domains, Bax beta has a unique carboxyl terminus and does not contain a hydrophobic transmembrane domain. Bcl-2 is also expressed in different Isoforms. Bcl-2 beta differs in the 3' UTR and coding region compared to variant alpha. Bcl-2 beta is shorter (22 kDa) and has a distinct C-terminus compared to Bcl-2 alpha (26 kDa). BAX is a member of the BCL-2 family of proteins, which function as regulators of apoptosis. Overexpression of BAX functions to promote cell death. BAX can form homodimers and is also able to heterodimerize with other BCL-2 related proteins.
Specifications
| Bax Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Q07812, Q07813, Q63690 | |
| BAX | |
| KLH conjugated to peptides corresponding to amino acids 3-16 of murine Bax (CGSGEQLGSGGPTSS), amino acids 3-16 of human Bax (CGSGEQPRGGGPTSS), and amino acids 12-24 shared by Bax from both species (CGPTSSEQIMKTGA). | |
| 200 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG κ |
| Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| 6A7 | |
| Unconjugated | |
| BAX | |
| apoptosis regulator BAX; Apoptosis regulator Bcl-X; Bax; Bax zeta; BAX-ALPHA; Bax-alpha protein; Baxdelta2G9; Baxdelta2G9omega; Baxdelta2omega; BCL2 associated X protein; BCL2 associated X, apoptosis regulator; BCL2 like 1; BCL2-associated protein; Bcl-2-associated protein Bax; Bcl2-associated protein Bax; Bcl-2-associated X protein; BCL2-associated X protein; BCL2-associated X protein omega; BCL2L; BCL2L1; Bcl2-L-1; BCL2L4; Bcl2-L-4; BCL2-like 1; bcl-2-like protein 1; Bcl-2-like protein 4; BCLX; Bcl-X; BCL-XL/S; PPP1R52; protein phosphatase 1, regulatory subunit 52 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 12028, 24887, 581 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.