Learn More
Asecos™ BATTERY CHARGE Charging Cabinet ION-CLASSIC-90
Safe active storage of lithium-ion batteries with integrated 3-stage warning/fire suppression system
Brand:  Asecos™ 3727604738082
Additional Details : Weight : 527.00000kg
 View more versions of this product
Product Code. 16274561
  • 7791.00 € / Each
Estimated Shipment: 06-11-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Alert:
For indoor use only
Description
Description
  • Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Easy handling with comfort: smooth doors with permanent self-closing function and oil-dampened door closer; open doors with a minimum amout of force
  • No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
  • Easy transport: integrated transport base with removable, optional base cover
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated technical ventilation to avoid heat build-up, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
  • All-around protection: 90 minute fire protection from outside to inside (type 90 / type tested in accordance with EN 14470-1) and for more than 90 minutes fire resistance for fires from inside to outside
  • Safe storage and charging: installed 3-stage warning/fire suppression system including smoke detector, temperature sensor, visual and acoustic alarms and fire suppression unit; triggers automatically in case of a fire; plug-in ready for connection to mains supply
Specifications
Specifications
EU-version
Safe active storage of lithium-ion batteries
Sheet Steel
615 x 1193 x 2224 mm
Anthracite Gray, Gentian Blue
180 kg
IO90.195.120.K3.WDC
2
Charging cabinet
Powder-coated textured
Bottom collecting sump (sheet steel powder-coated) 33 L capacity
522 x 1050 x 1647 mm
Grid
Wing
5
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, CE compliant