missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAT1 Mouse anti-Human, Mouse, Clone:3A2B2, Abnova™
Mouse monoclonal antibody raised against recombinant human BAT1.
Brand: Abnova MAB17134.100ug
This item is not returnable.
View return policy
Description
This gene encodes a member of the DEAD box family of RNA-dependent ATPases that mediate ATP hydrolysis during pre-mRNA splicing. The encoded protein is an essential splicing factor required for association of U2 small nuclear ribonucleoprotein with pre-mRNA, and also plays an important role in mRNA export from the nucleus to the cytoplasm. A cluster of genes, BAT1-BAT5, is localized in the vicinity of the genes for TNF alpha and TNF beta. These genes are all within the human major histocompatibility complex class III region. Mutations in this gene may be associated with rheumatoid arthritis. Alternatively spliced transcript variants encoding the same protein have been described. [provided by RefSeq]Specifications
| BAT1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunocytochemistry Flow Cytometry The optimal working dilution should be determined by the end user. |
|
| BAT1 | |
| Mouse | |
| 100 μg | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3A2B2 | |
| HLA-B associated transcript 1 | |
| In PBS (0.05% sodium azide) | |
| D6S81E, DDX39B, UAP56 | |
| Recombinant protein corresponding to amino acid 1-250 of human BAT1 from E. coli. | |
| 7919 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction