Learn More
Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories
Shop All Thermo Scientific ProductsDescription
Optimize your ultrapure water purification system with Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories. Increase flexibility in your lab with the remote dispenser, which delivers water up to 8' (2.4m) from the system and ships with a mounting bracket. Gain protection with the low pressure pump protector which alerts the E-Pure of an inadequate feed water condition in a pressurized feed line. The low water level pump protector alerts the system of an inadequate feed water condition when fed from a reservoir.
Specifications
Specifications
| For Use With (Equipment) | Barnstead E-Pure |
| Type | Remote Dispenser |
| Voltage | 0/1 V |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.