Learn More
Thermo Scientific™ Barnstead™ Cartridge Kits
Product Code. 11746259 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Nanopure II, Older Nanopure Systems
Nanopure Infinity Systems
Nanopure Quad-style System
Feedwater Source:
Deionized, Tap
Reverse Osmosis, Distilled
Tap
Tap or Pretreated
Model:
D3804
D3805
D4801
D4802
D50254
D50255
D50256
D50257
Unit Size:
Each
Product Code. 11746259
Brand: Thermo Scientific™ D4801
Only null left
Add to basket
Add to basket
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Tap
For Use With (Equipment) Nanopure Quad-style System
Includes One 09-034 high capacity ion exchange, 09-034-104 Organic/colloid and two 09-050-276 Ultrapure™ cartridges
Model D4801
Type Water Purification Cartridge