missing translation for 'onlineSavingsMsg'
Learn More

Barium trifluoromethanesulfonate, Thermo Scientific™

Product Code. 11348877
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348877 10 g 10g
11338877 2 g 2g
11358877 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11348877 Supplier Thermo Scientific Alfa Aesar Supplier No. 038571.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2794-60-7
Molecular Formula C2BaF6O6S2
Molecular Weight (g/mol) 435.453
MDL Number MFCD00143644
InChI Key DXJURUJRANOYMX-UHFFFAOYSA-L
Synonym barium trifluoromethanesulfonate, barium triflate, methanesulfonic acid, trifluoro-, barium salt, barium trifluoromethanesulphonate, methanesulfonic acid, 1,1,1-trifluoro-, barium salt 2:1, bariumtrifluoromethanesulfonate, pubchem23759, ksc491a8b, barium 2+ ; trifluoromethanesulfonate
PubChem CID 76040
IUPAC Name barium(2+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Ba+2]

Specifications

Melting Point >300°C
Quantity 10 g
UN Number UN1564
Beilstein 3729346
Sensitivity Hygroscopic
Formula Weight 435.48
Physical Form Powder
Chemical Name or Material Barium trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.