missing translation for 'onlineSavingsMsg'
Learn More

BAR501, MedChemExpress

Product Code. 16463318
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463318 10 mg 10mg
16486037 5 mg 5mg
16454566 100 mg 100mg
16440749 1 mg 1mg
16439856 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16463318 Supplier Medchem Express Supplier No. HY101274
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress BAR501 is a potent and selective agonist of GPBAR1 with an EC50 of 1 μM.

BAR501 is a potent and selective agonist of GPBAR1 with an EC50 of 1 μM.

Specifications

Chemical Name or Material BAR501
CAS 1632118-69-4
Color White
Purity Grade Notes Research
Molecular Formula C26H46O3
Quantity 10 mg
Solubility Information Ethanol : 120 mg/mL (295.10 mM; Need ultrasonic) ∣DMSO : ≥ 50 mg/mL (122.96 mM)
SMILES C[C@H](CCCO)[C@@]1([H])CC[C@@]2([H])[C@]3([H])[C@@H](O)[C@@H](CC)[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])CC[C@@]21C
Molecular Weight (g/mol) 406.64
Formula Weight 406.64
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.