missing translation for 'onlineSavingsMsg'
Learn More

BAMB-4, MedChemExpress

Product Code. 16437138
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16437138

Brand: Medchem Express HY16694

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BAMB-4(ITPKA-IN-C14) is a new membrane-permeable inhibitor against inositol-1,4,5-trisphosphate-3-kinase A((ITPKA) with IC50 of 37 μM in ADP-Glo Assay.

BAMB-4(ITPKA-IN-C14) is a new membrane-permeable inhibitor against inositol-1,4,5-trisphosphate-3-kinase A((ITPKA) with IC50 of 37 μM in ADP-Glo Assay.

Specifications

Chemical Name or Material BAMB-4
Physical Form Solid
CAS 891025-25-5
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C15H12N2O2
For Use With (Application) Cancer-Kinase/protease
Synonym ITPKA-IN-C14
Solubility Information DMSO : ≥ 40 mg/mL (158.56 mM)
SMILES O=C(C1=CC=C(C)C=C1)NC2=NOC3=CC=CC=C23
Molecular Weight (g/mol) 252.27
Formula Weight 252.27
Percent Purity 99.46%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.