Learn More
Balofloxacin, MedChemExpress
Product Code. 16478136
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 16478136
Brand: Medchem Express HYB0159
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Balofloxacin (Q-35) is an orally active fluoroquinolone antibiotic with broad-spectrum antibacterial activity against gram-negative, gram-positive, and anaerobic bacteria. Balofloxacin can be used for the research of respiratory, intestinal, and urinary tract infections.
Balofloxacin (Q-35) is an orally active fluoroquinolone antibiotic with broad-spectrum antibacterial activity against gram-negative, gram-positive, and anaerobic bacteria. Balofloxacin can be used for the research of respiratory, intestinal, and urinary tract infections.
Specifications
Chemical Name or Material Balofloxacin
Physical Form Solid
CAS 127294-70-6
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H24FN3O4
For Use With (Application) COVID-19-immunoregulation
Synonym Q-35
Solubility Information H2O : 0.67 mg/mL (1.72 mM; Need ultrasonic) ∣DMSO : 0.67 mg/mL (1.72 mM; Need ultrasonic)
SMILES O=C(C1=CN(C2CC2)C3=C(C=C(F)C(N4CC(NC)CCC4)=C3OC)C1=O)O
Molecular Weight (g/mol) 389.42
Formula Weight 389.42
Percent Purity 99.37%
Grade Research