Learn More
behr Labor-Technik™ Ballon Canister
Product Code. 10024710
Change view
Click to view available options
Capacity (Metric):
20 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10024710 20 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10024710 Supplier behr Labor-Technik™ Supplier No. B00176996
  • ONLINE EXCLUSIVE PRICE
    186.90 €
    144.88 € / Pack of 1
    Save 42.02 €
    22% Off
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The blue behroplast™ canisters are light fast. Your level display made of polyurethane ensures additional usage options. Naturally, they also have the welded tap for maximum tightness and optimum usage value.
  • Canister
  • blue with light protection
  • level indicator and tap
  • Simple transport of the behrotest™ canister
  • Space-saving injected handle
  • Easy removal of liquids
  • Very easy removal with the welded behrotest™tap
  • Practical dimensions
  • Space-saving storage
  • Level display made of polyurethane
  • The liquid level directly in view
  • Individually packed canisters
  • Protection against damage and soiling
Specifications
Product Type Ballon Canister
Capacity (Metric) 20 L
Color Blue
Material LDPE
Includes Tap and Level Indicator
Length (Metric) 280 mm
Width (Metric) 250 mm
Height (Metric) 370 mm