missing translation for 'onlineSavingsMsg'
Learn More

Balicatib, MedChemExpress

Product Code. 16436636
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16436636

Brand: Medchem Express HY15100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Balicatib (AAE581) is a potent, orally active and selective cathepsin K inhibitor with IC50 values of 22, 61, 48, 2900 nM for cathepsin K, cathepsin B, cathepsin L, cathepsin S, respectively. Balicatib inhibits bone turnover, decreases bone formation rates. Balicatib has the potential for the research of osteoporosis.

Balicatib (AAE581) is a potent, orally active and selective cathepsin K inhibitor with IC50 values of 22, 61, 48, 2900 nM for cathepsin K, cathepsin B, cathepsin L, cathepsin S, respectively. Balicatib inhibits bone turnover, decreases bone formation rates. Balicatib has the potential for the research of osteoporosis.

Specifications

Chemical Name or Material Balicatib
Physical Form Solid
CAS 354813-19-7
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C23H33N5O2
For Use With (Application) Neuroscience-Neuromodulation
Synonym AAE581
Solubility Information DMSO : 75 mg/mL (182.24 mM; Need ultrasonic)
SMILES CCCN1CCN(C2=CC=C(C(NC3(C(NCC#N)=O)CCCCC3)=O)C=C2)CC1
Molecular Weight (g/mol) 411.54
Formula Weight 411.54
Percent Purity 99.07%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.