missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BAGE3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17909322
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17909322 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17909322 Supplier Invitrogen™ Supplier No. PA5144148

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BAGE3 Polyclonal Antibody for IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen BAGE3
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene BAGE3
Gene Accession No. Q86Y29
Gene Alias B melanoma antigen 3; B melanoma antigen family member 3; B melanoma antigen family, member 3; BAGE3; Cancer/testis antigen 2.3; cancer/testis antigen family 2, member 3; CT2.3
Gene Symbols BAGE3
Host Species Rabbit
Immunogen Recombinant Human B melanoma antigen 3 protein (18-109AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85318
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.