missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bag-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
214.00 € - 301.00 €
Specifications
| Antigen | Bag-1 |
|---|---|
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18060886
|
Novus Biologicals
NB100-56082SS |
0.025 mL |
214.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18036254
|
Novus Biologicals
NB100-56082 |
0.05 mL |
301.00 €
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bag-1 Polyclonal specifically detects Bag-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Bag-1 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Tumor Suppressors | |
| Whole antisera with 0.02% Sodium Azide | |
| 573 | |
| A synthetic peptide corresponding to amino acids 26-45 (NERYDLLVTPQQGNSEPVVQD) of mouse BAG-1 was used as immunogen; GenBank no. AAH03722.1. This sequence is 100% conserved in mouse BAG-1L; GenBank no. NP_033866.3. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat | |
| BAG family molecular chaperone regulator 1, BAG-1, Bcl-2 associating athanogene-1 protein, BCL2-associated athanogene, Bcl-2-associated athanogene 1, Bcl-2-binding protein, glucocortoid receptor-associated protein RAP46,46-KD, HAP, RAP46 | |
| BAG1 | |
| IgG | |
| Unpurified | |
| BAG1 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title