Learn More
PeproTech BAFF Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Goat Polyclonal Antibody
219.00 € - 3435.00 €
Specifications
| Antigen | BAFF |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17834503
|
PeproTech
500-P163GBT-25UG |
25 μg |
219.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17824063
|
PeproTech
500-P163GBT-50UG |
50 μg |
275.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17804513
|
Invitrogen
500-P163GBT-1MG |
2 x 500 μg | |||||||
|
30054489
|
PeproTech
500-P163GBT-500UG |
500 μg |
1836.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: AVQGPEETVT QDCLQLIADS ETPTIQKGSY TFVPWLLSFK RGSALEEKEN KILVKETGYF FIYGQVLYTD KTYAMGHLIQ RKKVHVFGDE LSLVTLFRCI QNMPETLPNN SCYSAGIAKL EEGDELQLAI PRENAQISLD GDVTFFGALK LL. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human BAFF. Anti-Human BAFF-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hBAFF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human BAFF (500-P163G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hBAFF. Western Blot: To detect hBAFF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hBAFF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P163GBT-1MG will be provided as 2 x 500 μg
BAFF is a membrane bound cytokine expressed by B cell lineage cells. The soluble form of the cytokine binds to the receptors TNFRSF13B/TACI, TNFRSF17/BCMA, and TNFRSF13C/BAFFR. BAFF is a potent B cell activator, and important for the proliferation and differentiation of B cells. BAFF is also expressed by T lymphocytes and has been shown to promote T cell activation and survival. Isoforms representing alternatively spliced, and glycosylated variants have been identified.Specifications
| BAFF | |
| ELISA, Western Blot | |
| Biotin | |
| Goat | |
| Human | |
| Q9Y275 | |
| 10673 | |
| E.coli-derived Recombinant Human BAFF. | |
| Antigen affinity chromatography | |
| TNFSF13B |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| ApoL related ligand TALL-1; b cell-activating factor; B lymphocyte stimulator; BAFF; B-cell-activating factor; B-lymphocyte stimulator; BLYS; CD257; D8Ertd387e; delta BAFF; Delta4 BAFF; dendritic cell-derived TNF-like molecule; DTL; RGD1561519; TALL1; TALL-1; THANK; TNF- and APOL-related leukocyte expressed ligand 1; TNF and ApoL-related leukocyte expressed ligand 1; TNF homolog that activates apoptosis; Tnfsf13b; TNFSF20; TNLG7A; tumor necrosis factor (ligand) superfamily member 13b; tumor necrosis factor (ligand) superfamily, member 13b; tumor necrosis factor (ligand) superfamily, member 20; tumor necrosis factor ligand 7A; tumor necrosis factor ligand superfamily member 13B; Tumor necrosis factor ligand superfamily member 13b, membrane form; Tumor necrosis factor ligand superfamily member 13b, soluble form; tumor necrosis factor superfamily member 13b; tumor necrosis factor-like protein ZTNF4; UNQ401/PRO738; ZTNF4 | |
| TNFSF13B | |
| Primary | |
| -20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.