missing translation for 'onlineSavingsMsg'
Learn More

Bafetinib, MedChemExpress

Product Code. 16488118
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16488118

Brand: Medchem Express HY50868

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bafetinib is a potent and orally active Lyn/Bcr-Abl tyrosine kinase inhibitor. Bafetinib augments the activities of several proapoptotic Bcl-2 homology (BH)3-only proteins (Bim, Bad, Bmf and Bik) and induces apoptosis in Ph+ leukemia cells via Bcl-2 family-regulated intrinsic apoptosis pathway.

Bafetinib is a potent and orally active Lyn/Bcr-Abl tyrosine kinase inhibitor. Bafetinib augments the activities of several proapoptotic Bcl-2 homology (BH)3-only proteins (Bim, Bad, Bmf and Bik) and induces apoptosis in Ph+ leukemia cells via Bcl-2 family-regulated intrinsic apoptosis pathway.

Specifications

Chemical Name or Material Bafetinib
Physical Form Solid
CAS 859212-16-1
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C30H31F3N8O
For Use With (Application) Cancer-Kinase/protease
Synonym INNO-406 NS-187
Solubility Information DMSO : 100 mg/mL (173.42 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C(C(F)(F)F)=C1)CN2C[C@@H](N(C)C)CC2)NC3=CC=C(C)C(NC4=NC=CC(C5=CN=CN=C5)=N4)=C3
Molecular Weight (g/mol) 576.62
Formula Weight 576.62
Percent Purity 99.18%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.