Learn More
Thermo Scientific™ Geobacillus stearothermophilus ATCC™ 7953™
Click to view available options
ATCC®:
7953TM*
Description:
Spore Strip Bacillus stearothermophilus ATCC™ 7953™; 105 spore population
Spore Strip Bacillus stearothermophilus ATCC™ 7953™; 106spore population
Unit Size:
100 strips
Description
Thermo Scientific™ Spore Strips are stabilized microorganism-impregnated strips packaged in individual recloseable plastic bags.
- 10 -6spore population
Specifications
Specifications
| ATCC® | 7953TM* |
| Description | Spore Strip Bacillus stearothermophilus ATCC™ 7953™; 106spore population |
| Format | Spore Strips |
| Product Type | Geobacillus Species |
| Target Organism Class | Gram-positive Rods |
| Test Type | Qualitative QC |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.