Learn More
BOLA B.safe Canister GL45
Description
- Funnel made of PE / static dissipative PE-EX for liquid waste disposal
- The funnel can be safely mounted on the waste container with its freely turnable cap that is safely fixed with the filling neck.
- b.safe Funnels 180 are available in many versions
- The sieve restrains magnetic stirrer bars and other solid materials and can be easily removed for cleaning
- The hinged lid respectively the ball valve reliably withhold vapours from the container
- The splash guard avoids spillover and splashing during funnel filling
- The static dissipative PE-EX version includes two attachable filling tubes
- The mounted filling tube allows an under level filling to avoid static charging
Specifications
Specifications
| Product Type | Safety Funnel |