Learn More
Usbeck Laborgeraete Brush, D=90mm
Brand:  Usbeck Laborgeraete 5052
Brush, D=90mm
Product Code. 10005661
  • ONLINE EXCLUSIVE PRICE
    8.45 €
    7.40 € / Each
    Save 1.05 €
    12% Off
Only null left
Estimated Shipment: 12-02-2026