missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B-D-Threo-thymidylyl-(3'→5')-B-D-threo-thymidine-13D2 Ammonia Salt, TRC
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Specifications
Specifications
| Chemical Name or Material | B-D-Threo-thymidylyl-(3'-5')-B-D-threo-thymidine-13C2 Ammonia Salt |
| Molecular Formula | C1813C2H27N4O12P.NH3 |
| Quantity | 5 mg |
| Synonym | ((2R,3S,5R)-3-Hydroxy-5-(5-(methyl-13C)-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-2-yl)methyl ((2R,3S,5R)-2-(hydroxymethyl)-5-(5-(methyl-13C)-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-3-yl) hydrogen phosphate ammonia |
| InChI Formula | InChI=1S/C20H27N4O12P.H3N/c1-9-5-23(19(29)21-17(9)27)15-3-11(26)14(35-15)8-33-37(31,32)36-12-4-16(34-13(12)7-25)24-6-10(2)18(28)22-20(24)30;/h5-6,11-16,25-26H,3-4,7-8H2,1-2H3,(H,31,32)(H,21,27,29)(H,22,28,30);1H3/t11-,12-,13+,14+,15+,16+;/m0./s1 |
| SMILES | O=C(N([C@@H]1O[C@H](CO)[C@@H](OP(OC[C@H]([C@@H](O)C2)O[C@H]2N(C(N3)=O)C=C(C)C3=O)(O)=O)C1)C=C4C)NC4=O.N |
| IUPAC Name | ((2R,3S,5R)-3-Hydroxy-5-(5-(methyl-13C)-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-2-yl)methyl ((2R,3S,5R)-2-(hydroxymethyl)-5-(5-(methyl-13C)-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-3-yl) hydrogen phosphate ammonia |
| Molecular Weight (g/mol) | 565.44 |
| Formula Weight | 565.17 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction