missing translation for 'onlineSavingsMsg'
Learn More

b-AP15, MedChemExpress

Product Code. p-200098361
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
25mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16493396

Brand: Medchem Express HY13989

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress b-AP15 is a specific inhibitor of the deubiquitinating enzymes UCHL5 and Usp14.

b-AP15 is a specific inhibitor of the deubiquitinating enzymes UCHL5 and Usp14.

Specifications

Chemical Name or Material b-AP15
CAS 1009817-63-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H17N3O6
Quantity 25 mg
Synonym NSC 687852
Solubility Information DMSO : 20 mg/mL (47.69 mM; Need ultrasonic)
SMILES O=C1/C(CN(C(C=C)=O)C/C1=C\C2=CC=C([N+]([O-])=O)C=C2)=C/C3=CC=C([N+]([O-])=O)C=C3
Molecular Weight (g/mol) 419.39
Formula Weight 419.39
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.