Learn More
Brady™ B-423 Polyester Thermal Transfer Labels
Product Code. 16677865
Click to view available options
Height (Metric):
3.2 mm
4.8 mm
5.1 mm
6.4 mm
9.5 mm
9.7 mm
10.2 mm
11.1 mm
12.7 mm
19.1 mm
31.8 mm
Labels per Row:
1
2
3
4
5
7
12
Vertical Repeat (Metric):
6.4 mm
7.3 mm
7.6 mm
8.9 mm
9.5 mm
12.1 mm
12.7 mm
13.6 mm
15.2 mm
22.2 mm
34.9 mm
Width (Metric):
6.4 mm
9.5 mm
12.7 mm
16.5 mm
17.8 mm
19.1 mm
22.9 mm
25.4 mm
27.9 mm
31.8 mm
34.9 mm
38.1 mm
50.8 mm
69.9 mm
Unit Size:
1 roll
Each
Product Code. 16677865
Brand: Brady™ 030357
Please to purchase this item. Need a web account? Register with us today!
Printable, polyester labels for Thermal Transfer Printers.
  • Permanent adhesive
  • For smooth surfaces
Specifications
Autoclavable Autoclavable
Certifications/Compliance UL, CSA and AGA Approved
Adhesive Type Permanent Acrylic
Color White
Finish Gloss
Format Die-cut
Material Polyester (B-423)
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Barcode Labeling, Circuit Board Labeling, Equipment Labeling, General Identification, Inventory Labeling
For Use With (Equipment) BBP81;i5100;i7100;PR Plus;IP Series;THT 76 mm Core Printers;i7500
Labels per Row 1
Min. Operating Temperature (Metric) -70°C
Model THT-79-423-10
No. per Pack 10000 Label(s) / Roll
Quantity 10,000 Labels
Shape Rectangle
Size 50.80 mm (W) x 10.16 mm (H)
Temperature Range (English) -94°F to +230°F
Temperature Range (Metric) -70°C to +110°C
Thickness (English) 0.003 in
Thickness (Metric) 0.076 mm
Vertical Repeat (Metric) 12.7 mm
Dimensions (LxW) 50.80 mm (W) x 10.16 mm (H)
Width (English) 2 in
Width (Metric) 50.8 mm
Width (English) Printable Area 50.80 mm
Width (Metric) Printable Area 50.8 mm
Height (English) 0.4 in
Height (Metric) 10.2 mm
Height (Metric) Printable Area 10.2 mm
Alternate Part Number THT-79-423-10