Learn More
Azomethine-H, TRC
Specifications
Specifications
| CAS | 32266-60-7 |
| Molecular Formula | C17 H13 N O8 S2 |
| Quantity | 5 g |
| Synonym | 2,7-Naphthalenedisulfonic acid, 4-hydroxy-5-[[(2-hydroxyphenyl)methylene]amino]- (9CI, ACI), 1-Naphthol-3,6-disulfonic acid, 8-salicylideneamino- (6CI), 2,7-Naphthalenedisulfonic acid, 4-hydroxy-5-(salicylideneamino)- (8CI), 4-Hydroxy-5-[[(2-hydroxyphenyl)methylene]amino]-2,7-naphthalenedisulfonic acid (ACI), Azomethine H, NCI 88947 |
| InChI Formula | InChI=1S/C17H13NO8S2/c19-15-4-2-1-3-10(15)9-18-14-7-12(27(21,22)23)5-11-6-13(28(24,25)26)8-16(20)17(11)14/h1-9,19-20H,(H,21,22,23)(H,24,25,26)/b18-9+ |
| SMILES | Oc1ccccc1\C=N\c2cc(cc3cc(cc(O)c23)S(=O)(=O)O)S(=O)(=O)O |
| IUPAC Name | 4-hydroxy-5-[(E)-(2-hydroxyphenyl)methylideneamino]naphthalene-2,7-disulfonic acid |
| Molecular Weight (g/mol) | 423.42 |
| Formula Weight | 423.0083 |
| Chemical Name or Material | Azomethine-H |