missing translation for 'onlineSavingsMsg'
Learn More

Azobenzene, MedChemExpress

Product Code. p-200099378
Encompass_Preferred
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 16448738

Brand: Medchem Express HYB2127

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Azobenzene can be used as an optical trigger for the design and synthesis of a large variety of photoresponsive systems.

Azobenzene can be used as an optical trigger for the design and synthesis of a large variety of photoresponsive systems.

Specifications

Chemical Name or Material Azobenzene
CAS 103-33-3
Color Orange
Purity Grade Notes Research
Molecular Formula C12H10N2
Quantity 10 g
Solubility Information DMSO : ≥ 150 mg/mL (823.18 mM)
SMILES C1(/N=N/C2=CC=CC=C2)=CC=CC=C1
Molecular Weight (g/mol) 182.22
Formula Weight 182.22
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.