Learn More
Azlon™ Polypropylene and PVC Hand Pump
Description
- Liquid is forced into the collecting vessel by squeezing the bulb
- The flow can be instantly interrupted by pressing the release button
- Can be used as a siphon by attaching tubing to the outlet tube and after pumping the liquid will flow continuously
Specifications
Specifications
| Quantity | 1 Pack |
| Type | Hand Pump |
| For Use With (Application) | Can be used with various sizes of bottles due to telescopic down tube |
| Material | Polypropylene, PVC |
| Diameter (Metric) Outer Stopper | 44 x 60 mm |