Learn More
Azlon™ Polypropylene and PVC Hand Pump
Product Code. 10146354
Change view
Click to view available options
:
Outer Stopper Diameter: 44 x 60mm; Flow Rate: 12L/min.; Length: 70mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10146354 Outer Stopper Diameter: 44 x 60mm; Flow Rate: 12L/min.; Length: 70mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10146354 Supplier Azlon Supplier No. PWJ016
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used with various sizes of bottles due to telescopic down tube
  • Liquid is forced into the collecting vessel by squeezing the bulb
  • The flow can be instantly interrupted by pressing the release button
  • Can be used as a siphon by attaching tubing to the outlet tube and after pumping the liquid will flow continuously
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Pack
Type Hand Pump
For Use With (Application) Can be used with various sizes of bottles due to telescopic down tube
Material Polypropylene, PVC
Diameter (Metric) Outer Stopper 44 x 60 mm