Learn More
Azinphos-ethyl-d10 (diethyl-d10), CDN
Specifications
Specifications
| Chemical Name or Material | Azinphos-ethyl D10 (ethyl D10) |
| CAS | 2732916-08-2 |
| Molecular Formula | C12 2H10 H6 N3 O3 P S2 |
| Quantity | 0.01 g |
| Synonym | Azinphos-ethyl D10 (ethyl D10) |
| InChI Formula | InChI=1 S/C12H16N3O3PS2/c1-3-17-19(20,18-4-2)21-9-15-12(16)10-7-5-6-8-11(10)13-14-15/h5-8 H,3-4,9H2,1-2H3/i1D3,2D3,3D2,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])OP(=S)(OC([2 H])([2 H])C([2 H])([2 H])[2 H])SCN1N=Nc2ccccc2C1=O |
| IUPAC Name | 3-[bis(1,1,2,2,2-pentadeuterioethoxy)phosphinothioylsulfanylmethyl]-1,2,3-benzotriazin-4-one |
| Molecular Weight (g/mol) | 355.44 |
| Formula Weight | 355.0998 g/mol |
| Show More |