Learn More
Azido-PEG3-carboxyrhodamine 110 conjugate, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mg
25 mg
Description
It is suitable for confocal laser scanning microscopy. Its long PEG spacer should reduce any steric effect of the dye on the molecule to be labeled. Carboxyrhodamine 110 and its derivatives are superior alternatives to other green fluorescent dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is suitable for confocal laser scanning microscopy. Its long PEG spacer should reduce any steric effect of the dye on the molecule to be labeled. Carboxyrhodamine 110 and its derivatives are superior alternatives to other green fluorescent dyes.
Solubility
Soluble in DMSO, DMF or methanol.
Notes
Excite using the 488nm spectral line of the argon ion laser
It is suitable for confocal laser scanning microscopy. Its long PEG spacer should reduce any steric effect of the dye on the molecule to be labeled. Carboxyrhodamine 110 and its derivatives are superior alternatives to other green fluorescent dyes.
Solubility
Soluble in DMSO, DMF or methanol.
Notes
Excite using the 488nm spectral line of the argon ion laser
Specifications
Color | Red |
Quantity | 25 mg |
Solubility Information | Soluble in DMSO,DMF or methanol. |
Formula Weight | 574.59 |
Physical Form | Solid |
Chemical Name or Material | Azido-PEG3-carboxyrhodamine 110 conjugate |