Learn More
AZD3229 Tosylate, MedChemExpress
Product Code. 16493206
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493206 25 mg 25mg
16470997 50 mg 50mg
16464708 10 mg 10mg
16445536 100 mg 100mg
16407427 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16493206 Supplier Medchem Express Supplier No. HY112802A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress AZD3229 Tosylate is a potent pan-KIT mutant inhibitor for the treatment of gastrointestinal stromal tumors.
AZD3229 Tosylate is a potent pan-KIT mutant inhibitor for the treatment of gastrointestinal stromal tumors.
Specifications
Chemical Name or Material AZD3229 Tosylate
CAS 2248003-71-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C31H34FN7O6S
Quantity 25 mg
Solubility Information DMSO : ≥ 100 mg/mL (153.44 mM)
SMILES COCCOC1=CC(F)=C2C(N=CN=C2NC3=CC=C(NC(CN4C=C(C(C)C)N=N4)=O)C=C3)=C1.CC5=CC=C(S(O)(=O)=O)C=C5
Molecular Weight (g/mol) 651.71
Formula Weight 651.71
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease