Learn More
AZD3229, MedChemExpress
Product Code. 16497417
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16497417
Brand: Medchem Express HY112802
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress AZD3229 is a potent pan-KIT mutant inhibitor for the treatment of gastrointestinal stromal tumors.
AZD3229 is a potent pan-KIT mutant inhibitor for the treatment of gastrointestinal stromal tumors.
Specifications
Chemical Name or Material AZD3229
CAS 2248003-60-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H26FN7O3
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (208.55 mM)
SMILES COCCOC1=CC(F)=C2C(N=CN=C2NC3=CC=C(NC(CN4C=C(C(C)C)N=N4)=O)C=C3)=C1
Molecular Weight (g/mol) 479.51
Formula Weight 479.51
Percent Purity 99.83%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease