missing translation for 'onlineSavingsMsg'
Learn More

AZD2906, MedChemExpress

Product Code. 16411057
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16411057

Brand: Medchem Express HY113854

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZD2906 is a selective glucocorticoid receptor (GR) agonist, increases micronucleated immature erythrocytes in the bone marrow of rats. AZD2906 shows IC50s of 2.2, 0.3, 41.6 and 7.5 nM at GR in human, rat PBMC and human, rat whole blood, respectively.

AZD2906 is a selective glucocorticoid receptor (GR) agonist, increases micronucleated immature erythrocytes in the bone marrow of rats. AZD2906 shows IC50s of 2.2, 0.3, 41.6 and 7.5 nM at GR in human, rat PBMC and human, rat whole blood, respectively.

Specifications

Chemical Name or Material AZD2906
CAS 1034148-15-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C26H25FN4O3
Quantity 50 mg
Solubility Information DMSO : 125 mg/mL (271.44 mM; Need ultrasonic)
SMILES FC1=CC=C(N2N=CC3=CC(O[C@@H]([C@@H](NC(C4CC4)=O)C)C5=CC=C(N=C5)OC)=CC=C23)C=C1
Molecular Weight (g/mol) 460.5
Formula Weight 460.5
Percent Purity 99.82%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.