missing translation for 'onlineSavingsMsg'
Learn More

AZD0156, MedChemExpress

Product Code. 16424206
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16424206 100 mg 100mg
16425487 5 mg 5mg
16409446 50 mg 50mg
16402748 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16424206 Supplier Medchem Express Supplier No. HY100016

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZD0156 is a potent, selective and orally active ATM inhibitor with an IC50 of 0.58 nM. AZD0156 inhibits the ATM-mediated signaling, prevents DNA damage checkpoint activation, disrupts DNA damage repair, and induces tumor cell apoptosis.

AZD0156 is a potent, selective and orally active ATM inhibitor with an IC50 of 0.58 nM. AZD0156 inhibits the ATM-mediated signaling, prevents DNA damage checkpoint activation, disrupts DNA damage repair, and induces tumor cell apoptosis.

Specifications

Chemical Name or Material AZD0156
CAS 1821428-35-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H31N5O3
Quantity 100 mg
Solubility Information DMSO : 20 mg/mL (43.33 mM; ultrasonic and adjust pH to 3 with HCl)
SMILES O=C(N1C2CCOCC2)N(C)C3=C1C4=CC(C5=CC=C(OCCCN(C)C)N=C5)=CC=C4N=C3
Molecular Weight (g/mol) 461.56
Formula Weight 461.56
Percent Purity 99.62%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.