Learn More
Tocris Bioscience™ AZD 9272
Product Code. 18791578
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18791578 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18791578 Supplier Tocris Bioscience™ Supplier No. 5613/50
Please to purchase this item. Need a web account? Register with us today!
Potent and selective mGlu5 antagonist; brain penetrant
AZD 9272 is a potent and selective mGlu5 antagonist (IC50 values are 2.6 and 7.6 nM for rat and human receptors, respectively). Exhibits >3900-fold selectivity for mGlu5 over other mGlu receptors. Also selective over a panel of other targets. Displays discriminative effects in rats. Orally bioavailable and brain penetrant.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Fluoro-5-[3-(5-fluoro-2-pyridinyl)-1,2,4-oxadiazol-5-yl]benzonitrile
CAS 327056-26-8
Quantity 50 mg
Target Group I mGluR Antagonists
Molecular Formula C14H6F2N4O
Formula Weight 284.22
Purity >98%