missing translation for 'onlineSavingsMsg'
Learn More

AZD-3463, MedChemExpress

Product Code. p-200101698
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489237 5 mg 5mg
16436836 100 mg 100mg
16422767 50 mg 50mg
16416688 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16489237 Supplier Medchem Express Supplier No. HY15609

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZD-3463 (ALK/IGF1R inhibitor) is an orally active ALK/IGF1R inhibitor, with a Ki of 0.75 nM for ALK. AZD3463 induces apoptosis and autophagy in neuroblastoma cells.

AZD-3463 (ALK/IGF1R inhibitor) is an orally active ALK/IGF1R inhibitor, with a Ki of 0.75 nM for ALK. AZD3463 induces apoptosis and autophagy in neuroblastoma cells.

Specifications

Chemical Name or Material AZD-3463
CAS 1356962-20-3
Color Green
Purity Grade Notes Research
Molecular Formula C24H25ClN6O
Quantity 5 mg
Synonym ALK/IGF1R inhibitor
Solubility Information DMSO : 20 mg/mL (44.55 mM; Need ultrasonic)
SMILES ClC(C=NC(NC1=C(OC)C=C(N2CCC(N)CC2)C=C1)=N3)=C3C4=CNC5=C4C=CC=C5
Molecular Weight (g/mol) 448.95
Formula Weight 448.95
Percent Purity 98.27%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.