Learn More
Azaphen dihydrochloride monohydrate, MedChemExpress
Description
Pipofezine(Azafen or Azaphen) is a potent inhibitor of the reuptake of serotonin.
Specifications
Specifications
| Chemical Name or Material | Azaphen dihydrochloride monohydrate |
| CAS | 63302-99-8 |
| Color | Yellow-Green |
| Purity Grade Notes | Research |
| Molecular Formula | C16H23Cl2N5O2 |
| Quantity | 500 mg |
| Synonym | Azafen dihydrochloride monohydrate Pipofezin dihydrochloride monohydrate Pipofezine dihydrochloride monohydrate Azaphenonxazine dihydrochloride monohydrate |
| Solubility Information | H2O : ≥ 100 mg/mL (257.54 mM) ∣DMSO : 1 mg/mL (2.58 mM; Need ultrasonic) |
| SMILES | [H]Cl.[H]Cl.CN1C2=CC=CC=C2OC3=C1C=C(N4CCN(C)CC4)N=N3.O |
| Molecular Weight (g/mol) | 388.29 |
| Show More |