missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azaphen dihydrochloride monohydrate, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Pipofezine(Azafen or Azaphen) is a potent inhibitor of the reuptake of serotonin.
Specifications
Specifications
| Chemical Name or Material | Azaphen dihydrochloride monohydrate |
| CAS | 63302-99-8 |
| Color | Yellow-Green |
| Purity Grade Notes | Research |
| Molecular Formula | C16H23Cl2N5O2 |
| Quantity | 100 mg |
| Synonym | Azafen dihydrochloride monohydrate Pipofezin dihydrochloride monohydrate Pipofezine dihydrochloride monohydrate Azaphenonxazine dihydrochloride monohydrate |
| Solubility Information | H2O : ≥ 100 mg/mL (257.54 mM) ∣DMSO : 1 mg/mL (2.58 mM; Need ultrasonic) |
| SMILES | [H]Cl.[H]Cl.CN1C2=CC=CC=C2OC3=C1C=C(N4CCN(C)CC4)N=N3.O |
| Molecular Weight (g/mol) | 388.29 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction