Learn More
AZ1495, MedChemExpress
Product Code. 16495296
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495296 100 mg 100mg
16493056 25 mg 25mg
16437127 5 mg 5mg
16424408 10 mg 10mg
16420727 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16495296 Supplier Medchem Express Supplier No. HY111101
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress AZ1495, a weak base, is a potent orally active interleukin-1 receptor associated kinase 4 (IRAK4) inhibitor. AZ1495 has a favorable physicochemical and kinase selectivity for IRAK4 and IRAK1 with IC50 values of 0.005 μM and 0.023 μM, respectively. AZ1495 has IRAK4 inhibition with a Kd value of 0.0007 μM. AZ1495 can be used for the research of diffuse large B-cell lymphoma (DLBCL).
AZ1495, a weak base, is a potent orally active interleukin-1 receptor associated kinase 4 (IRAK4) inhibitor. AZ1495 has a favorable physicochemical and kinase selectivity for IRAK4 and IRAK1 with IC50 values of 0.005 μM and 0.023 μM, respectively. AZ1495 has IRAK4 inhibition with a Kd value of 0.0007 μM. AZ1495 can be used for the research of diffuse large B-cell lymphoma (DLBCL).
Specifications
Chemical Name or Material AZ1495
Physical Form Solid
CAS 2196204-23-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H31N5O2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 10 mg/mL (25.94 mM; Need ultrasonic)
SMILES C1(C(C2CCOCC2)=CN3)=C3N=CN=C1N[C@@H]4CC[C@@H](N5CCOCC5)CC4
Molecular Weight (g/mol) 385.5
Formula Weight 385.5
Percent Purity 98.0%
Grade Research