missing translation for 'onlineSavingsMsg'
Learn More

AZ1495, MedChemExpress

Product Code. 16424408
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16424408 10 mg 10mg
16495296 100 mg 100mg
16493056 25 mg 25mg
16437127 5 mg 5mg
16420727 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16424408 Supplier Medchem Express Supplier No. HY111101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZ1495, a weak base, is a potent orally active interleukin-1 receptor associated kinase 4 (IRAK4) inhibitor. AZ1495 has a favorable physicochemical and kinase selectivity for IRAK4 and IRAK1 with IC50 values of 0.005 μM and 0.023 μM, respectively. AZ1495 has IRAK4 inhibition with a Kd value of 0.0007 μM. AZ1495 can be used for the research of diffuse large B-cell lymphoma (DLBCL).

AZ1495, a weak base, is a potent orally active interleukin-1 receptor associated kinase 4 (IRAK4) inhibitor. AZ1495 has a favorable physicochemical and kinase selectivity for IRAK4 and IRAK1 with IC50 values of 0.005 μM and 0.023 μM, respectively. AZ1495 has IRAK4 inhibition with a Kd value of 0.0007 μM. AZ1495 can be used for the research of diffuse large B-cell lymphoma (DLBCL).

Specifications

Chemical Name or Material AZ1495
Physical Form Solid
CAS 2196204-23-4
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C21H31N5O2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 10 mg/mL (25.94 mM; Need ultrasonic)
SMILES C1(C(C2CCOCC2)=CN3)=C3N=CN=C1N[C@@H]4CC[C@@H](N5CCOCC5)CC4
Molecular Weight (g/mol) 385.5
Formula Weight 385.5
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.