Learn More
Tocris Bioscience™ AZ 10606120 dihydrochloride
GREENER_CHOICE
Product Code. 18074772
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18074772
Brand: Tocris Bioscience™ 3323/50
Please to purchase this item. Need a web account? Register with us today!
Potent P2X7 receptor antagonist
Potent P2X7 receptor antagonist (KD values are 1.4 and 19 nM at human and rat P2X7 receptors respectively). Binds in a positive cooperative manner to sites distinct from, but coupled to, the ATP binding site and acts as a negative allosteric modulator. Inhibits tumor growth and displays antiangiogenic effects in mice.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 607378-18-7
Molecular Formula C25H36Cl2N4O2
Specifications
Quantity 50 mg
Formula Weight 495.48
Purity >98%
Molecular Weight (g/mol) 495.489
Chemical Name or Material AZ 10606120 dihydrochloride