Learn More
AZ 10606120 Dihydrochloride (~90%), TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Specifications
Specifications
| Chemical Name or Material | AZ 10606120 Dihydrochloride |
| CAS | 607378-18-7 |
| Molecular Formula | C25H34N4O2 • 2(HCl) |
| Quantity | 2.5 mg |
| Synonym | N-[2-[[2-[(2-Hydroxyethyl)amino]ethyl]amino]-5-quinolinyl]-2-tricyclo[3.3.1.13,7]dec-1-ylacetamide Dihydrochloride, N-[2-[[2-[(2-Hydroxyethyl)amino]ethyl]amino]-5-quinolinyl]-2-tricyclo[3.3.1.13,7]dec-1-ylacetamide Dihydrochloride (9CI), 2-(1-Adamantyl)-N-[2-[[2-[(2-hydroxyethyl)amino]ethyl]amino]quinolin-5-yl]acetamide Dihydrochloride |
| InChI Formula | InChI=1S/C25H34N4O2.2ClH/c30-9-8-26-6-7-27-23-5-4-20-21(28-23)2-1-3-22(20)29-24(31)16-25-13-17-10-18(14-25)12-19(11-17)15-25;;/h1-5,17-19,26,30H,6-16H2,(H,27,28)(H,29,31);2*1H |
| SMILES | O=C(CC12CC(C3)CC(C2)CC3C1)NC4=C5C=CC(NCCNCCO)=NC5=CC=C4 |
| IUPAC Name | 2-(1-adamantyl)-N-[2-[2-(2-hydroxyethylamino)ethylamino]quinolin-5-yl]acetamide;dihydrochloride |
| Molecular Weight (g/mol) | 422.5623646 |