Learn More
Axygen™ Presterilized Conical Centrifuge Tubes
GREENER_CHOICE
Product Code. 12539107 Shop All Axygen Scientific Products
Click to view available options
Capacity (Metric):
15 mL
50 mL
Packaging:
25 Tubes and Caps/Rack, 20 Racks/Cs.
25 Tubes and Caps/Unit, 20 Units/Cs.
50 Tubes and Caps/Unit, 20 Units/Cs.
Bag
Unit Size:
Pack of 500
Product Code. 12539107
Brand: Axygen™ SCT50MLRS
Please to purchase this item. Need a web account? Register with us today!
Graduated tubes with conical bottom
  • Rated to 12,000 RCF
  • Printed graduations and writing area for sample identification
  • Screw caps can be closed with one hand.
  • Sterile options are also available.
  • RNase-/DNase-free
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY
Specifications
Cap Style Screw Cap
Closure Material HDPE
Autoclavable Not Autoclavable
Capacity (Metric) 50 mL
Color Clear
Material Polypropylene
For Use With (Application) For sample identification
Includes Screw cap
No. per Pack 250/Pk.
Packaging 25 Tubes and Caps/Rack, 20 Racks/Cs.
Sterility Sterile
Graduated Yes
Closure Type Screw Cap
Bottom Shape Cylindrical
Max. RCF 12,000 x g
Product Type Centrifuge Tube