Learn More
Axygen™ Polypropylene Round Bottom 96 Well Assay Plates
Shop All Axygen Scientific Products
Click to view available options
Color:
Clear
Unit Size:
Pack of 50
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Color | Clear |
| Material | Polypropylene |
| No. of Wells | 96 |
| Total Volume (Metric) Well | 2 mL |
| Well Shape | Round Bottom (U-well) |
| For Use With (Application) | Sample Collection |