Learn More
Axygen™ 384-well Skirted PCR Microplates
Product Code. 12769438 Shop All Axygen Scientific Products
Click to view available options
Color:
Blue
Clear
Green
Orange
Red
Violet
White
Yellow
Includes:
Double notch
Single notch
Unit Size:
Pack of 100
Pack of 20
Pack of 50
Volume (Metric) Well:
40 μL
50 μL
55 μL
Product Code. 12769438
Brand: Axygen™ PCR384Y
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Single and double notch 384-well PCR plates
Sealing Options: Axymats, heat sealing, adhesive films and foils.
  • Available in 50μL and 55μL
  • Single and double notch corner
Skirted (50μL)
  • Application/Compatibility: standard thermal cycling
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted (55μL)
  • Application/Compatibility: standard thermal cycling, fully ABI compatible for qPCR and sequencing
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted, RigidPlate (50μL)
  • Robotic-friendly rigid plate with full skirt
  • Application/Compatibility: standard thermal cycling, ABI sequencing, automation applications
  • Packaging Description: 10 plates/Pk., 2 packs/sleeve, 5 sleeves/Cs.
Skirted, elevated wells
  • Application/Compatibility: standard thermal cycling, ABI sequencing, qPCR
  • Raised wells allow 55μL volume
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Colors available by special order.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RNase, DNase-free
Format Standard
Array 16 x 24
Color Yellow
Material Polypropylene
No. of Wells 384
Volume (Metric) Working 40 μL
Sterility Non-sterile
Well Shape Round
For Use With (Application) PCR
Includes Double notch
Well Color Yellow